(4-Bromobutyl)triphenylphosphonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23040-09
B23040-18
CAAAB23040-09EA
62.09
CAD
CAAAB23040-09
CAAAB23040-18
(4-Bromobutyl)triphenylphosphonium bromide 98%
(4-Bromobutyl)triphenylphosphonium bromide
Purity: 98%
CAS No.: 7333-63-3
Molecular Formula: Br(CH2)4P(C6H5)3Br
Formula Weight: 478.22
Melting Point: 206-212°
Beilstein Reference: 3812291
Formula:
C₂₂H₂₃Br₂P MW: 478.21 g/mol Melting Pt: 206…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031636 CAS Number: 7333-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...