3-Tolylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23025-03
B23025-06
B23025-14
CAAAB23025-03EA
71.57
CAD
CAAAB23025-03
CAAAB23025-06
CAAAB23025-14
3-Tolylboronic acid 97%
3-Tolylboronic acid
Suzuki coupling reactions
Purity: 97%
CAS No.: 17933-03-8
Molecular Formula: C7H9BO2
Formula Weight: 135.96
Melting Point: ca 160-162°
Beilstein Reference: 2935968
Formula:
C₇H₉BO₂ Melting Pt: ∼ 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040198 CAS Number: 17933-03-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...