Methyl-2-nitrobenzoate 98+%

Supplier: Thermo Scientific Chemicals

B23009-14 B23009-22
CAAAB23009-14EA 164.32 CAD
CAAAB23009-14 CAAAB23009-22 CAAAB23009-36
Methyl-2-nitrobenzoate 98+%
Methyl 2-nitrobenzoate

Purity: 98+% CAS No.: 606-27-9 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Flash Point: 110°(230°F) Refractive Index: 1.5330 Melting Point: -13° Boiling Point: 275° Beilstein Reference: 2050101 MDL No.: MFCD00007136

Formula: C₈H₇NO₄
Boiling Pt: 275 °C
Melting Pt: –13 °C
Density: 1.285
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007136
CAS Number: 606-27-9
EINECS: 210-111-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR