2-(Hydroxymethyl)benzyl alcohol 97%
Supplier: Thermo Scientific Chemicals
B23008-03
B23008-06
B23008-14
CAAAB23008-03EA
74.58
CAD
CAAAB23008-03
CAAAB23008-06
CAAAB23008-14
2-(Hydroxymethyl)benzyl alcohol 97%
2-(Hydroxymethyl)benzyl alcohol
Purity: 97%
CAS No.: 612-14-6
Molecular Formula: C8H10O2
Formula Weight: 138.17
Melting Point: 59-64°
Beilstein Reference: 508775
Formula:
C₈H₁₀O₂ Boiling Pt: 291 °C (1013 hPa) Melting Pt: 59…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004626 CAS Number: 612-14-6 EINECS: 210-293-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...