tert-Butyl ethyl ether 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22945-06
B22945-14
B22945-22
CAAAB22945-06EA
113.29
CAD
CAAAB22945-06
CAAAB22945-14
CAAAB22945-22
tert-Butyl ethyl ether 99%
tert-Butyl ethyl ether
Purity: 99%
CAS No.: 637-92-3
Molecular Formula: (CH3)3COCH2CH3
Formula Weight: 102.18
Flash Point: -19°(-2°F)
Refractive Index: 1.3750
Melting Point: -94°
Boiling Point: 72-73°
Beilstein Reference: 1731469
Merck Reference: 14,3774
UN No.: 1179
MDL No.: MFCD00009225
Formula:
C₆H₁₄O Boiling Pt: 72…73 °C Melting Pt: –94 °C Density: 0.750 Flash Pt: –19 °C (–2 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009225 CAS Number: 637-92-3 EINECS: 211-309-7 UN: 1179 ADR: 3,II Merck Index: 14,3774 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...