Pentafluorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3,4,5,6-Pentafluorobenzeneboronic acid, Dihydroxy(pentafluorophenyl)borane, (Pentafluorophenyl)metaboric acid, Pentafluorobenzeneboronic acid

B22922-06 B22922-14
CAAAB22922-06EA 170.64 CAD
CAAAB22922-06 CAAAB22922-14
Pentafluorophenylboronic acid 97%
Pentafluorophenylboronic acid
Formula: C₆H₂BF₅O₂
MW: 211.88 g/mol
Melting Pt: 250…255 °C
Storage Temperature: Ambient
MDL Number: MFCD01074663
CAS Number: 1582-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR