3-(Benzyloxy)benzaldehyde 98%

Supplier: Thermo Scientific Chemicals

B22909-06 B22909-14 B22909-22
CAAAB22909-06EA 61.15 CAD
CAAAB22909-06 CAAAB22909-14 CAAAB22909-22
3-(Benzyloxy)benzaldehyde 98%
3-(Benzyloxy)benzaldehyde

Purity: 98% CAS No.: 1700-37-4 Molecular Formula: C14H12O2 Formula Weight: 212.25 Melting Point: 54-58° Boiling Point: 180°/0.08mm MDL No.: MFCD00003367

Formula: C₁₄H₁₂O₂
Boiling Pt: 180 °C/0.08 mm
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003367
CAS Number: 1700-37-4
EINECS: 216-932-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR