1,12-Dodecanediol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,12-Dihydroxy dodecane

B22885-14 B22885-22
CAAAB22885-14EA 92.59 CAD
CAAAB22885-14 CAAAB22885-22
1,12-Dodecanediol 99%
1,12-Dodecanediol

Purity: 99% CAS No.: 5675-51-4 Molecular Formula: HO(CH2)12OH Formula Weight: 202.34 Melting Point: 81-84° Boiling Point: 188-190°/12mm MDL No.: MFCD00004755

Formula: C₁₂H₂₆O₂
MW: 202.34 g/mol
Boiling Pt: 188…190 °C/12 mm
Melting Pt: 81…84 °C
Density: 1.08 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004755
CAS Number: 5675-51-4
EINECS: 227-133-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR