(2-Chloro-4-fluorophenyl)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(2-Chloro-4-fluorophenyl)acetic acid

B22872-06 B22872-14
CAAAB22872-06EA 94.33 CAD
CAAAB22872-06 CAAAB22872-14
(2-Chloro-4-fluorophenyl)acetic acid 97%
(2-Chloro-4-fluorophenyl)acetic acid

Purity: 97% CAS No.: 177985-32-9 Molecular Formula: C8H6ClFO2 Formula Weight: 188.59 Melting Point: 102-105° MDL No.: MFCD00236028

Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59
Boiling Pt: 289
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00236028
CAS Number: 177985-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR