1,4-Diisopropylbenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22871-14
B22871-22
B22871-36
CAAAB22871-14EA
69.84
CAD
CAAAB22871-14
CAAAB22871-22
CAAAB22871-36
1,4-Diisopropylbenzene 99%
1,4-Diisopropylbenzene
Purity: 99%
CAS No.: 100-18-5
Molecular Formula: C12H18
Formula Weight: 162.27
Flash Point: 76°(170°F)
Refractive Index: 1.4890
Odor: Aromatic
Melting Point: -17°
Boiling Point: 210°
Beilstein Reference: 1854739
MDL No.: MFCD00008892
Formula:
C₁₂H₁₈ Boiling Pt: 210 °C Melting Pt: –17 °C Density: 0.857 Flash Pt: 76 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008892 CAS Number: 100-18-5 EINECS: 202-826-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...