1-Bromo-2,4-dinitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Danger

B22867-14 B22867-22
CAAAB22867-14EA 145.36 CAD
CAAAB22867-14 CAAAB22867-22
1-Bromo-2,4-dinitrobenzene 98%
1-Bromo-2,4-dinitrobenzene

Purity: 98% CAS No.: 584-48-5 Molecular Formula: C6H3BrN2O4 Formula Weight: 247.01 Melting Point: 70-73° UN No.: 3459 MDL No.: MFCD00041873

Formula: C₆H₃BrN₂O₄
Melting Pt: 70…73 °C
Density: Min. 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041873
CAS Number: 584-48-5
EINECS: 209-539-8
UN: 2928
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR