Cloxiquina 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-8-quinolinol, 5-Chloroquinolin-8-ol, 5-Chloro-8-hydroxyquinoline

B22856-18 B22856-30
CAAAB22856-18EA 62.57 CAD
CAAAB22856-18 CAAAB22856-30
Cloxiquina 95%
Cloxiquina

Purity: 95% CAS No.: 130-16-5 Molecular Formula: C9H6ClNO Formula Weight: 179.61 Melting Point: 122-124° Beilstein Reference: 5289 Merck Reference: 14,2422 MDL No.: MFCD00006788

Formula: C₉H₆ClNO
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00006788
CAS Number: 130-16-5
EINECS: 204-978-1
Merck Index: 14,2422

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR