N,N,N-Trimethylanilinium chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethylphenylammonium chloride, Phenyltrimethylammonium chloride
B22851-22
B22851-36
CAAAB22851-22EA
75.21
CAD
CAAAB22851-22
CAAAB22851-36
N,N,N-Trimethylanilinium chloride 98+%
N,N,N-Trimethylanilinium chloride
Purity: 98+%
CAS No.: 138-24-9
Molecular Formula: C6H5N(CH3)3Cl
Formula Weight: 171.67
Melting Point: ca 236° subl.
Beilstein Reference: 3572527
UN No.: 2811
MDL No.: MFCD00011790
Formula:
C₉H₁₄ClN Melting Pt: ∼ 236 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00011790 CAS Number: 138-24-9 EINECS: 205-319-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...