(±)-Bromosuccinic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22829-06
B22829-14
B22829-22
CAAAB22829-06EA
76
CAD
CAAAB22829-06
CAAAB22829-14
CAAAB22829-22
(±)-Bromosuccinic acid 96%
(±)-Bromosuccinic acid
Purity: 96%
CAS No.: 923-06-8
Molecular Formula: C4H5BrO4
Formula Weight: 196.99
Melting Point: 164-166° dec.
Beilstein Reference: 1723556
Merck Reference: 14,1437
MDL No.: MFCD00004213
Formula:
HOOCBrCHCH₂COOH Boiling Pt: 319 °C (1013 hPa) Melting Pt: 164…166 °C (dec.) Density: 2.073 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004213 CAS Number: 923-06-8 EINECS: 213-087-7 Merck Index: 14,1437 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...