3,4-Difluorophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B22799-03 B22799-06 B22799-14
CAAAB22799-03EA 22.28 CAD
CAAAB22799-03 CAAAB22799-06 CAAAB22799-14
3,4-Difluorophenylboronic acid 98%
3,4-Difluorophenylboronic acid

Purity: 98% CAS No.: 168267-41-2 Molecular Formula: C6H5BF2O2 Formula Weight: 157.91 Melting Point: 300° Beilstein Reference: 7369788

Formula: C₆H₅BF₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00807405
CAS Number: 168267-41-2
EINECS: 605-510-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR