2,3-Dichlorophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B22781-03 B22781-06 B22781-14
CAAAB22781-03EA 57.83 CAD
CAAAB22781-03 CAAAB22781-06 CAAAB22781-14
2,3-Dichlorophenylboronic acid 98%
2,3-Dichlorophenylboronic acid

Purity: 98% CAS No.: 151169-74-3 Molecular Formula: C6H5BCl2O2 Formula Weight: 190.82 Melting Point: 233-242° MDL No.: MFCD01075703

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 233…242 °C
Storage Temperature: Ambient
MDL Number: MFCD01075703
CAS Number: 151169-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR