Benzylacetone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22777-0B
B22777-30
CAAAB22777-0BEA
243.32
CAD
CAAAB22777-0B
CAAAB22777-30
Benzylacetone 98%
Benzylacetone
Purity: 98%
CAS No.: 2550-26-7
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 105°(221°F)
Refractive Index: 1.5122
Boiling Point: 235-237°
Beilstein Reference: 1907123
MDL No.: MFCD00008790
Formula:
(C₆H₅)CH₂CH₂C(O)CH₃ Boiling Pt: 235…237 °C Melting Pt: –13 °C Density: 0.988 Flash Pt: 105 °C(221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008790 CAS Number: 2550-26-7 EINECS: 219-847-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...