2-Aminofluorene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22769-03
B22769-06
B22769-14
CAAAB22769-03EA
33.02
CAD
CAAAB22769-03
CAAAB22769-06
CAAAB22769-14
2-Aminofluorene 98%
2-Aminofluorene
Purity: 98%
CAS No.: 153-78-6
Molecular Formula: C13H11N
Formula Weight: 181.24
Melting Point: 126-130°
Beilstein Reference: 1945861
Formula:
C₁₃H₁₁N Boiling Pt: 327 °C (1013 hPa) Melting Pt: 126…130 °C Density: 1.281 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001125 CAS Number: 153-78-6 EINECS: 205-817-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...