3,5-Dichlorophenylboronic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22765-03
B22765-06
CAAAB22765-03EA
60.2
CAD
CAAAB22765-03
CAAAB22765-06
CAAAB22765-14
3,5-Dichlorophenylboronic acid 98+%
3,5-Dichlorophenylboronic acid
Purity: 98+%
CAS No.: 67492-50-6
Molecular Formula: C6H5BCl2O2
Formula Weight: 190.82
Melting Point: ca 315° dec.
Beilstein Reference: 7581091
Formula:
C₆H₅BCl₂O₂ Melting Pt: ∼ 315 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051935 CAS Number: 67492-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...