3-Fluoro-o-toluidine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22764-06
B22764-14
CAAAB22764-06EA
44.4
CAD
CAAAB22764-06
CAAAB22764-14
3-Fluoro-o-toluidine 98+%
3-Fluoro-o-toluidine
Purity: 98+%
CAS No.: 443-86-7
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 86°(186°F)
Refractive Index: 1.5420
Odor: Characteristic
Melting Point: 7°
Boiling Point: 205°
UN No.: 2810
MDL No.: MFCD00007760
Formula:
C₇H₈FN Boiling Pt: 205 °C Melting Pt: 7 °C Density: 1.099 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007760 CAS Number: 443-86-7 EINECS: 207-142-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...