3-Chloro-4-fluorophenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22755-06
B22755-14
CAAAB22755-03EA
76.77
CAD
CAAAB22755-03
CAAAB22755-06
CAAAB22755-14
3-Chloro-4-fluorophenylboronic acid 98%
3-Chloro-4-fluorophenylboronic acid
Purity: 98%
CAS No.: 144432-85-9
Molecular Formula: C6H5BClFO2
Formula Weight: 174.37
Melting Point: 255-260°
Beilstein Reference: 7369789
Formula:
C₆H₅BClFO₂ Melting Pt: 255…260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051800 CAS Number: 144432-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...