Diethyl pyrocarbonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

B22753-06 B22753-14 B22753-22
CAAAB22753-06EA 57.99 CAD
CAAAB22753-06 CAAAB22753-14 CAAAB22753-22
Diethyl pyrocarbonate 97%
Diethyl pyrocarbonate

Ribonuclease inhibitor


Purity: 97% CAS No.: 1609-47-8 Molecular Formula: C6H10O5 Formula Weight: 162.14 Storage Temperature: 2°C to 8°C Flash Point: 69°(156°F) Physical Form: Clear liquid Density: 1.101 Refractive Index: 1.3980 Sensitivity: Moisture Sensitive Appearance: Colorless Boiling Point: 93-94°/18mm Beilstein Reference: 637031 Merck Reference: 14,7998 MDL No.: MFCD00009106


Ribonuclease inhibitor

Formula: O(COOC₂H₅)₂
Boiling Pt: 93…94 °C/18 mm
Density: 1.101
Flash Pt: 69 °C(156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 14,7998

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR