5-Methoxyisatin 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22742-06
B22742-14
B22742-22
CAAAB22742-06EA
109.34
CAD
CAAAB22742-06
CAAAB22742-14
CAAAB22742-22
5-Methoxyisatin 98%
5-Methoxyisatin
Purity: 98%
CAS No.: 39755-95-8
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Melting Point: 199-202°
MDL No.: MFCD00169023
Formula:
C₉H₇NO₃ Melting Pt: 199…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00169023 CAS Number: 39755-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...