3,4-Dimethylanisole 99%
Supplier: Thermo Scientific Chemicals
Warning
|
B22735-18
B22735-30
CAAAB22735-18EA
104.6
CAD
CAAAB22735-18
CAAAB22735-30
3,4-Dimethylanisole 99%
3,4-Dimethylanisole
Purity: 99%
CAS No.: 4685-47-6
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 75°(167°F)
Refractive Index: 1.5175
Boiling Point: 200-201°
Beilstein Reference: 2040907
MDL No.: MFCD00008396
Formula:
C₉H₁₂O Boiling Pt: 200…201 °C Density: 0.975 Flash Pt: 75 °C(167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008396 CAS Number: 4685-47-6 EINECS: 225-142-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...