4,5-Dibromo-o-xylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2-Dibromo-4,5-dimethylbenzene, 4,5-Dibromo-1,2-dimethylbenzene
B22727-03
B22727-06
CAAAB22727-03EA
73.79
CAD
CAAAB22727-03
CAAAB22727-06
4,5-Dibromo-o-xylene 97%
4,5-Dibromo-o-xylene
Purity: 97%
CAS No.: 24932-48-7
Molecular Formula: C8H8Br2
Formula Weight: 263.97
Melting Point: 85-87°
Beilstein Reference: 2206183
Formula:
C₈H₈Br₂ Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082743 CAS Number: 24932-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...