2-Chloro-6-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

B22714-06 B22714-14
CAAAB22714-06EA 71.57 CAD
CAAAB22714-06 CAAAB22714-14
2-Chloro-6-(trifluoromethyl)aniline 97%
2-Chloro-6-(trifluoromethyl)aniline

Purity: 97% CAS No.: 433-94-3 Molecular Formula: C7H5ClF3N Formula Weight: 195.57 Boiling Point: 39-40°/0.1mm UN No.: 2810 MDL No.: MFCD00272565

Formula: C₇H₅ClF₃N
Boiling Pt: 39…40 °C/0.1 mm
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00272565
CAS Number: 433-94-3
EINECS: 207-091-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR