2-Amino-5-iodopyrimidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22705-03
B22705-06
B22705-14
CAAAB22705-03EA
56.25
CAD
CAAAB22705-03
CAAAB22705-06
CAAAB22705-14
2-Amino-5-iodopyrimidine 97%
2-Amino-5-iodopyrimidine
Purity: 97%
CAS No.: 1445-39-2
Molecular Formula: C4H4IN3
Formula Weight: 221.00
Melting Point: 217-220°
MDL No.: MFCD01075666
Formula:
C₄H₄IN₃ Melting Pt: 217…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075666 CAS Number: 1445-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...