2-Iodotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Iodo-2-methylbenzene

B22697-14 B22697-22 B22697-36
CAAAB22697-14EA 69.99 CAD
CAAAB22697-14 CAAAB22697-22 CAAAB22697-36
2-Iodotoluene 98%
2-Iodotoluene

Purity: 98% CAS No.: 615-37-2 Molecular Formula: C7H7I Formula Weight: 218.04 Flash Point: 90°(194°F) Refractive Index: 1.6090 Boiling Point: 210-212° Beilstein Reference: 1634294 MDL No.: MFCD00001042

Formula: C₇H₇I
MW: 218.04 g/mol
Boiling Pt: 210…212 °C
Density: 1.713
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001042
CAS Number: 615-37-2
EINECS: 210-422-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR