2-Iodo-4-(trifluoromethyl)aniline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22691-09
B22691-18
B22691-30
CAAAB22691-09EA
127.98
CAD
CAAAB22691-09
CAAAB22691-18
CAAAB22691-30
2-Iodo-4-(trifluoromethyl)aniline 99%
2-Iodo-4-(trifluoromethyl)aniline
Purity: 99%
CAS No.: 163444-17-5
Molecular Formula: C7H5F3IN
Formula Weight: 287.02
Melting Point: 49-52°
MDL No.: MFCD02683097
Formula:
C₇H₅F₃IN Melting Pt: 49…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683097 CAS Number: 163444-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...