(±)-3-Amino-1,2-propanediol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-3-Amino-1,2-dihydroxy propane, 3-Amino-1,2-propanediol

B22672-14 B22672-22 B22672-36
CAAAB22672-14EA 112.5 CAD
CAAAB22672-14 CAAAB22672-22 CAAAB22672-36
(±)-3-Amino-1,2-propanediol 97%
(±)-3-Amino-1,2-propanediol

Purity: 97% CAS No.: 616-30-8 Molecular Formula: C3H9NO2 Formula Weight: 91.11 Flash Point: 186°(366°F) Refractive Index: 1.4920 Boiling Point: 264-266°/739mm Beilstein Reference: 1719121 UN No.: 2735 MDL No.: MFCD00008140

Formula: C₃H₉NO₂
Boiling Pt: 264…266 °C/739 mm
Density: 1.186
Flash Pt: 186 °C (366 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008140
CAS Number: 616-30-8
EINECS: 210-475-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR