1,4-Diiodoperfluorobutane 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
B22660-06
B22660-14
CAAAB22660-06EA
135.72
CAD
CAAAB22660-06
CAAAB22660-14
1,4-Diiodoperfluorobutane 97% stabilized
1,4-Diiodoperfluorobutane
Purity: 97%
CAS No.: 375-50-8
Molecular Formula: C4F8I2
Formula Weight: 453.84
Refractive Index: 1.4294
Melting Point: -9°
Boiling Point: 85°/100mm
Beilstein Reference: 1777548
MDL No.: MFCD00042263
Formula:
C₄F₈I₂ Boiling Pt: 85 °C/100 mm Melting Pt: –9 °C Density: 2.487 Storage Temperature: Ambient |
MDL Number:
MFCD00042263 CAS Number: 375-50-8 EINECS: 206-788-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...