3',5'-Dimethoxy-4'-hydroxyacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 4'-Hydroxy-3',5'-dimethoxyacetophenone, 1-(3,5-Dimethoxy-4-hydroxyphenyl)ethan-1-one, Acetosyringone, 1-(4-Hydroxy-3,5-dimethoxyphenyl)ethanone
B22634-03
B22634-06
CAAAB22634-03EA
72.54
CAD
CAAAB22634-03
CAAAB22634-06
CAAAB22634-14
3',5'-Dimethoxy-4'-hydroxyacetophenone 97%
3',5'-Dimethoxy-4'-hydroxyacetophenone
Purity: 97%
CAS No.: 2478-38-8
Molecular Formula: C10H12O4
Formula Weight: 196.2
Melting Point: 117-122°
Beilstein Reference: 1966119
Formula:
C₁₀H₁₂O₄ Boiling Pt: 335 °C (1013 hPa) Melting Pt: 117…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008748 CAS Number: 2478-38-8 EINECS: 219-610-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...