3-Methyl-1,2-cyclohexanedione 98+%

Supplier: Thermo Scientific Chemicals

B22623-06 B22623-14
CAAAB22623-06EA 114.08 CAD
CAAAB22623-06 CAAAB22623-14
3-Methyl-1,2-cyclohexanedione 98+%
3-Methyl-1,2-cyclohexanedione

Purity: 98+% CAS No.: 3008-43-3 Molecular Formula: C7H10O2 Formula Weight: 126.16 Melting Point: 57-59° Boiling Point: 69-72°/1mm MDL No.: MFCD00209518

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 69…72 °C/1 mm
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00209518
CAS Number: 3008-43-3
EINECS: 221-122-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR