4-Isopropylphenol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22620-A1
B22620-18
B22620-30
CAAAAB22620-A1EA
364.98
CAD
CAAAAB22620-A1
CAAAB22620-18
CAAAB22620-30
4-Isopropylphenol 98%
4-Isopropylphenol
Purity: 98%
CAS No.: 99-89-8
Molecular Formula: C9H12O
Formula Weight: 136.20
Flash Point: 108°(226°F)
Melting Point: 59-61°
Boiling Point: 211-212°
Beilstein Reference: 1363564
UN No.: 2430
MDL No.: MFCD00002372
Formula:
C₉H₁₂O Boiling Pt: 211…212 °C Melting Pt: 59…61 °C Density: 0.99 g/cm³ (20 °C) Flash Pt: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002372 CAS Number: 99-89-8 EINECS: 202-798-8 UN: 2430 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...