4-Methoxybenzamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22614-09
B22614-18
CAAAB22614-09EA
81.37
CAD
CAAAB22614-09
CAAAB22614-18
4-Methoxybenzamide 98%
4-Methoxybenzamide
Purity: 98%
CAS No.: 3424-93-9
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 164-167°
Boiling Point: 295°
MDL No.: MFCD00007995
Formula:
C₈H₉NO₂ Boiling Pt: 295 °C Melting Pt: 164…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007995 CAS Number: 3424-93-9 EINECS: 222-319-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...