4-Heptylbenzoic acid 99+%
Supplier: Thermo Scientific Chemicals
B22589-06
B22589-14
CAAAB22589-06EA
98.12
CAD
CAAAB22589-06
CAAAB22589-14
4-Heptylbenzoic acid 99+%
4-Heptylbenzoic acid
Purity: 99+%
CAS No.: 38350-87-7
Molecular Formula: C14H20O2
Formula Weight: 220.31
Melting Point: 101-102°
Beilstein Reference: 2047135
Formula:
C₁₄H₂₀O₂ |
MDL Number:
MFCD00009722 CAS Number: 38350-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...