Dihexyl disulfide tech. 85%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

B22568-06 B22568-14
CAAAB22568-06EA 91.32 CAD
CAAAB22568-06 CAAAB22568-14
Dihexyl disulfide tech. 85%, Technical Grade
Dihexyl disulphide

Purity: 85% CAS No.: 10496-15-8 Molecular Formula: [CH3(CH2)5]2S2 Formula Weight: 234.47 Flash Point: 110°(230°F) Odor: Unpleasant MDL No.: MFCD00027300

Formula: C₁₂H₂₆S₂
Boiling Pt: 85 - 86 °C (0.5 -rr)
Density: 0.919
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027300
CAS Number: 10496-15-8
EINECS: 234-022-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR