Didecyl sulfide 97%

Supplier: Thermo Scientific Chemicals

B22560-09 B22560-18
CAAAB22560-09EA 47.24 CAD
CAAAB22560-09 CAAAB22560-18
Didecyl sulfide 97%
Didecyl sulphide

Purity: 97% CAS No.: 693-83-4 Molecular Formula: [CH3(CH2)9]2S Formula Weight: 314.63 Melting Point: 25-28° Boiling Point: 217-218°/8mm MDL No.: MFCD00026542

Formula: C₂₀H₄₂S
MW: 314.62 g/mol
Boiling Pt: 217…218 °C/8 mm
Melting Pt: 25…28 °C
Storage Temperature: Ambient
MDL Number: MFCD00026542
CAS Number: 693-83-4
EINECS: 211-761-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR