4-Methoxybenzyl mercaptan 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methoxy-α-toluenethiol, p-Methoxytoluene-α-thiol

B22542-06 B22542-14
CAAAB22542-06EA 101.59 CAD
CAAAB22542-06 CAAAB22542-14
4-Methoxybenzyl mercaptan 98%
4-Methoxybenzyl mercaptan

Purity: 98% CAS No.: 6258-60-2 Molecular Formula: C8H10OS Formula Weight: 154.23 Flash Point: 110°(230°F) Refractive Index: 1.5750 Odor: Unpleasant Boiling Point: 90-95°/0.5mm Beilstein Reference: 471686 MDL No.: MFCD00004871

Formula: C₈H₁₀OS
MW: 154.23
Boiling Pt: 90…95 °C/0.5 mm
Density: 1.112
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004871
CAS Number: 6258-60-2
EINECS: 228-393-6
UN: 3334
ADR: 9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR