5-Bromo-2-furaldehyde 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromofuran-2-carbaldehyde, 5-Bromofurfural
Stabilization:
Stabilised with ethanol 2%
B22536-09
B22536-18
CAAAB22536-09EA
151.68
CAD
CAAAB22536-09
CAAAB22536-18
5-Bromo-2-furaldehyde 98% stabilized
5-Bromo-2-furaldehyde
Purity: 98%
CAS No.: 1899-24-7
Molecular Formula: C5H3BrO2
Formula Weight: 174.98
Melting Point: 82-85°
Boiling Point: 112°/6mm
MDL No.: MFCD00159501
Formula:
C₅H₃BrO₂ MW: 174.98 g/mol Boiling Pt: 112 °C/6 mm Melting Pt: 82…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00159501 CAS Number: 1899-24-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...