(4-Hydroxyphenyl)acetic acid methyl ester 98+%

Supplier: Thermo Scientific Chemicals
Warning

B22533-14 B22533-22 B22533-36
CAAAB22533-14EA 78.84 CAD
CAAAB22533-14 CAAAB22533-22 CAAAB22533-36
(4-Hydroxyphenyl)acetic acid methyl ester 98+%
(4-Hydroxyphenyl)acetic acid methyl ester

Purity: 98+% CAS No.: 14199-15-6 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 55-58° Boiling Point: 163°/5mm MDL No.: MFCD00002387

Formula: C₉H₁₀O₃
Boiling Pt: 163 °C/5 mm
Melting Pt: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00002387
CAS Number: 14199-15-6
EINECS: 238-050-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR