(Methylthio)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Methylthio)ethansäure, 2-(Methylthio)acetic acid

B22531-03 B22531-06 B22531-14
CAAAB22531-03EA 59.57 CAD
CAAAB22531-03 CAAAB22531-06 CAAAB22531-14
(Methylthio)acetic acid 98%
(Methylthio)acetic acid

Purity: 98% CAS No.: 2444-37-3 Molecular Formula: CH3SCH2CO2H Formula Weight: 106.15 Flash Point: 110°(230°F) Refractive Index: 1.4950 Odor: Stench Melting Point: 13-14° Boiling Point: 99-100°/4mm Beilstein Reference: 1740201 MDL No.: MFCD00075444

Formula: C₃H₆O₂S
Boiling Pt: 99…100 °C/4 mm
Melting Pt: 13…14 °C
Density: 1.224
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075444
CAS Number: 2444-37-3
EINECS: 219-483-6
UN: 3334
ADR: "9",

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR