3-Amino-9-ethyl carbazole (AEC) 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22529-14
B22529-22
CAAAB22529-14EA
102.23
CAD
CAAAB22529-14
CAAAB22529-22
3-Amino-9-ethyl carbazole (AEC) 95%
3-Amino-9-ethyl carbazole
Peroxidase substrate for immunoblotting and immunohistochemical staining procedures
Purity: 95%
CAS No.: 132-32-1
Molecular Formula: C14H14N2
Formula Weight: 210.28
Odor: Odorless
Melting Point: 98-100°
MDL No.: MFCD00004964
Formula:
C₁₄H₁₄N₂ Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004964 CAS Number: 132-32-1 EINECS: 205-055-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...