3-Aminosalicylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22478-02
B22478-77
CAAAB22478-02EA
428.18
CAD
CAAAB22478-02
CAAAB22478-77
3-Aminosalicylic acid 97%
3-Aminosalicylic acid
Purity: 97%
CAS No.: 570-23-0
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Melting Point: ca 240° dec.
Beilstein Reference: 2090427
Formula:
C₇H₇NO₃ Boiling Pt: 347 °C (1013 hPa) Melting Pt: ∼ 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010299 CAS Number: 570-23-0 EINECS: 209-328-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...