4-Hydroxy-3,5-dinitrobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

B22474-03 B22474-06
CAAAB22474-03EA 98.59 CAD
CAAAB22474-03 CAAAB22474-06
4-Hydroxy-3,5-dinitrobenzoic acid 98+%
4-Hydroxy-3,5-dinitrobenzoic acid

Purity: 98+% CAS No.: 1019-52-9 Molecular Formula: C7H4N2O7 Formula Weight: 228.12 Melting Point: 248-252° MDL No.: MFCD00017000

Formula: C₇H₄N₂O₇
Melting Pt: 248…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00017000
CAS Number: 1019-52-9
EINECS: 213-814-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR