2-Toluenethiol 98%

Supplier: Thermo Scientific Chemicals
Danger

B22460-22 B22460-06 B22460-14
CAAAAB22460-22EA 673.08 CAD
CAAAAB22460-22 CAAAB22460-06 CAAAB22460-14
2-Toluenethiol 98%
2-Toluenethiol

Purity: 98% CAS No.: 137-06-4 Molecular Formula: C7H8S Formula Weight: 124.21 Flash Point: 63°(145°F) Refractive Index: 1.5800 Melting Point: 10-12° Boiling Point: 195° Beilstein Reference: 774068 Merck Reference: 14,9325 UN No.: 2810 MDL No.: MFCD00004838

Formula: C₇H₈S
Boiling Pt: 195 °C
Melting Pt: 10…12 °C
Density: 1.054
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004838
CAS Number: 137-06-4
EINECS: 205-276-8
UN: 3334
ADR: 6.1,III
Merck Index: 14,9325

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR