Hexadecylamine tech. 90% remainder mainly 1-octadecylamine, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22459-14
B22459-0B
B22459-30
CAAAAB22459-14EA
55.14
CAD
CAAAAB22459-14
CAAAB22459-0B
CAAAB22459-30
Hexadecylamine tech. 90% remainder mainly 1-octadecylamine, Technical Grade
Hexadecylamine
Purity: 90%
CAS No.: 143-27-1
Molecular Formula: CH3(CH2)15NH2
Formula Weight: 241.46
Flash Point: 140°(284°F)
Refractive Index: 1.4496
Melting Point: 42-47°
Boiling Point: 144°/2mm
Beilstein Reference: 1634065
UN No.: 3259
MDL No.: MFCD00008158
Formula:
H₃C(CH₂)₁₄CH₂NH₂ Boiling Pt: 144 °C/2 mm Melting Pt: 42…47 °C Density: 0.813 Flash Pt: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008158 CAS Number: 143-27-1 EINECS: 205-596-8 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...