2-Fluoro-4-iodoaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

B22450-06 B22450-14 B22450-22
CAAAB22450-06EA 60.99 CAD
CAAAB22450-06 CAAAB22450-14 CAAAB22450-22
2-Fluoro-4-iodoaniline 99%
2-Fluoro-4-iodoaniline

Purity: 99% CAS No.: 29632-74-4 Molecular Formula: C6H5FIN Formula Weight: 237.02 Flash Point: 110°(230°F) Melting Point: 55-57° Beilstein Reference: 2081100 MDL No.: MFCD00011738

Formula: C₆H₅FIN
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 55…57 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011738
CAS Number: 29632-74-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR