Sodium methanesulfonate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methanesulfonic acid sodium salt, Methanesulphonic acid sodium salt

B22439-06 B22439-14
CAAAB22439-06EA 66.83 CAD
CAAAB22439-06 CAAAB22439-14
Sodium methanesulfonate 98%
Sodium methanesulphonate

Purity: 98% CAS No.: 2386-57-4 Molecular Formula: CH3NaO3S Formula Weight: 118.09 Odor: Odorless

Formula: CH₃NaO₃S
MW: 118.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064389
CAS Number: 2386-57-4
EINECS: 219-203-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR