(±)-2-Phenylpropanal 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22436-22
B22436-36
CAAAB22436-22EA
79.79
CAD
CAAAB22436-22
CAAAB22436-36
(±)-2-Phenylpropanal 97%
(±)-2-Phenylpropanal
Purity: 97%
CAS No.: 93-53-8
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 79°(174°F)
Refractive Index: 1.5170
Boiling Point: 92-94°/12mm
Beilstein Reference: 4291321
MDL No.: MFCD00006973
Formula:
C₉H₁₀O Boiling Pt: 92…94 °C/12 mm Density: 1.010 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006973 CAS Number: 93-53-8 EINECS: 202-255-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...